ContactUsLinkComponent
This product was prepared from normal serum by delipidation, selective precipitation and tandem molecular sieve chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipi 1 * 1 mg
Catalog # AVIVOPRA03402
This product was prepared from normal serum by delipidation, selective precipitation and tandem molecular sieve chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipi 1 * 1 mg
Catalog # AVIVOPRA03402
Frequently Bought Together