Mouse Progranulin ELISA Kits
Catalog # ABCAAB213473-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Progranulin ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Progranulin ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Progranulin with a sensitivity of 7,8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.
Frequently Bought Together