ContactUsLinkComponent
ASTM-Thermom., S120C, white backed, 38,6+41,4:0,05C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.310x6-8mm,durable pigment, with ice point scale,government calibrated, with Calibration 1 * 1 items
Catalog # AMARA301610CB/CC3
Supplier:  Amarell
ASTM-Thermom., S120C, white backed, 38,6+41,4:0,05C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.310x6-8mm,durable pigment, with ice point scale,government calibrated, with Calibration 1 * 1 items
Catalog # AMARA301610CB/CC3
Supplier:  Amarell
Frequently Bought Together