ContactUsLinkComponent
Precision therm., similar ISO 655, long, +180+420:1C, enclosed scale, cap. colourless, gallium filled, total immersion, max. 375x9m m, richter-top finish with button, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN65092-GA
Supplier:  Amarell
Precision therm., similar ISO 655, long, +180+420:1C, enclosed scale, cap. colourless, gallium filled, total immersion, max. 375x9m m, richter-top finish with button, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN65092-GA
Supplier:  Amarell
Frequently Bought Together