ContactUsLinkComponent
Precision therm., similar ISO 655, long, -5+25:0,1C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max. 375x9mm, richter-top finish with button, null
Catalog # AMARN65002-CB
Supplier:  Amarell
Precision therm., similar ISO 655, long, -5+25:0,1C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max. 375x9mm, richter-top finish with button, null
Catalog # AMARN65002-CB
Supplier:  Amarell
Frequently Bought Together