ContactUsLinkComponent
Therm. similar Landsberger, +84+102:0,05C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid,, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63810-CB
Supplier:  Amarell
Therm. similar Landsberger, +84+102:0,05C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid,, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63810-CB
Supplier:  Amarell
Frequently Bought Together