ContactUsLinkComponent
Therm. similar Landsberger, +67+85:0,05C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special li quid, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63808-CB
Supplier:  Amarell
Therm. similar Landsberger, +67+85:0,05C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special li quid, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63808-CB
Supplier:  Amarell
Frequently Bought Together