ContactUsLinkComponent
Therm. similar Landsberger, +51+68:0,05C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special li quid, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63806-CB/CC
Supplier:  Amarell
Therm. similar Landsberger, +51+68:0,05C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special li quid, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63806-CB/CC
Supplier:  Amarell
Frequently Bought Together