ContactUsLinkComponent
Amarell-Special-Therm., +198+252:0,1C, enclosed scale, cap. round, colourless, gallium filled, c.450x9,0-9,5mm, Richter-top finish , with ice point scale, null
Catalog # AMARN61628-GA/CC
Supplier:  Amarell
Amarell-Special-Therm., +198+252:0,1C, enclosed scale, cap. round, colourless, gallium filled, c.450x9,0-9,5mm, Richter-top finish , with ice point scale, null
Catalog # AMARN61628-GA/CC
Supplier:  Amarell
Frequently Bought Together