ContactUsLinkComponent
Amarell-Special-Therm., -58+5:0,1C, enclosed scale, cap. round, colourless, toluene filled, c.450x9,0-9,5mm, Richter-top finish, su itable for government calibration 1 * 1 items
Catalog # AMARN61618-TOL
Supplier:  Amarell
Amarell-Special-Therm., -58+5:0,1C, enclosed scale, cap. round, colourless, toluene filled, c.450x9,0-9,5mm, Richter-top finish, su itable for government calibration 1 * 1 items
Catalog # AMARN61618-TOL
Supplier:  Amarell
Frequently Bought Together