ContactUsLinkComponent
Anschütztherm., similar DIN, +298+360:1°C, enclosed scale, cap. lens front, colourless, gallium filled, 175x5-6mm, total immersion, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARN60172-GA
Supplier:  Amarell
Anschütztherm., similar DIN, +298+360:1°C, enclosed scale, cap. lens front, colourless, gallium filled, 175x5-6mm, total immersion, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARN60172-GA
Supplier:  Amarell
Frequently Bought Together