ContactUsLinkComponent
Anschütztherm., similar DIN, +248+302:0,5C, enclosed scale, cap. lens front, colourless, gallium filled, 175x5-6mm, total immersio n, Richter-top finish, null
Catalog # AMARN60170-GA/VC
Supplier:  Amarell
Anschütztherm., similar DIN, +248+302:0,5C, enclosed scale, cap. lens front, colourless, gallium filled, 175x5-6mm, total immersio n, Richter-top finish, null
Catalog # AMARN60170-GA/VC
Supplier:  Amarell
Frequently Bought Together