ContactUsLinkComponent
Anschütztherm., similar DIN, +98+152:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid, 175x5-6mm, total immersion, Richter-top finish, null
  AMARN60164-CB/CC
 :  Amarell
Anschütztherm., similar DIN, +98+152:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid, 175x5-6mm, total immersion, Richter-top finish, null
  AMARN60164-CB/CC
 :  Amarell
Frequently Bought Together