ContactUsLinkComponent
Anschütztherm., similar DIN, +48+102:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid, 175x5-6mm, total immersion, Richter-top finish, null
Catalog # AMARN60162-CB
Supplier:  Amarell
Anschütztherm., similar DIN, +48+102:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid, 175x5-6mm, total immersion, Richter-top finish, null
Catalog # AMARN60162-CB
Supplier:  Amarell
Frequently Bought Together