ContactUsLinkComponent
Anschütztherm., similar DIN, 0+52:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liq uid, 175x5-6mm, total immersion, Richter-top finish, null
  AMARN60160-CB
 :  Amarell
Anschütztherm., similar DIN, 0+52:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liq uid, 175x5-6mm, total immersion, Richter-top finish, null
  AMARN60160-CB
 :  Amarell
Frequently Bought Together