ContactUsLinkComponent
Anschütztherm., +48+102:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liqu id, 340x5-6mm, total immersion, with button at top, null
Catalog # AMARN60074-CB
Supplier:  Amarell
Anschütztherm., +48+102:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liqu id, 340x5-6mm, total immersion, with button at top, null
Catalog # AMARN60074-CB
Supplier:  Amarell
Frequently Bought Together