ContactUsLinkComponent
Anschütz-Therm., +298+360:0,1°C, enclosed scale, cap. lens front, colourless, gallium filled, 340x5-6mm, Richter-top finish, suitabl e for government calibration 1 * 1 items
Catalog # AMARN60012-GA
Supplier:  Amarell
Anschütz-Therm., +298+360:0,1°C, enclosed scale, cap. lens front, colourless, gallium filled, 340x5-6mm, Richter-top finish, suitabl e for government calibration 1 * 1 items
Catalog # AMARN60012-GA
Supplier:  Amarell
Frequently Bought Together