ContactUsLinkComponent
Anschütz-Therm., -10+52:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, 340x5- 6mm, Richter-top finish, null
Catalog # AMARN60000-CB/CC
Supplier:  Amarell
Anschütz-Therm., -10+52:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, 340x5- 6mm, Richter-top finish, null
Catalog # AMARN60000-CB/CC
Supplier:  Amarell
Frequently Bought Together