ContactUsLinkComponent
Therm., similar Dr. Otte, similar DIN, +200+450:1C, enclosed scale, cap. lens front, colourless, galliumfilled, 370x7mm, 2954 glass , Richter-top finish, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN59806-GA
Supplier:  Amarell
Therm., similar Dr. Otte, similar DIN, +200+450:1C, enclosed scale, cap. lens front, colourless, galliumfilled, 370x7mm, 2954 glass , Richter-top finish, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN59806-GA
Supplier:  Amarell
Frequently Bought Together