ContactUsLinkComponent
Melting point therm., +100+250:0,5C, enclosed scale, cap. prismatic, colourless, gallium filled, 250x6-7mm, immersion 30mm, Richter -top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM250402-GA
Supplier:  Amarell
Melting point therm., +100+250:0,5C, enclosed scale, cap. prismatic, colourless, gallium filled, 250x6-7mm, immersion 30mm, Richter -top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM250402-GA
Supplier:  Amarell
Frequently Bought Together