ContactUsLinkComponent
Melting point therm., -38+100:0,5C, enclosed scale, cap. prismatic, colourless, red special liquid, 250x6-7mm, immersion 30mm, Rich ter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM250400-FL
Supplier:  Amarell
Melting point therm., -38+100:0,5C, enclosed scale, cap. prismatic, colourless, red special liquid, 250x6-7mm, immersion 30mm, Rich ter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM250400-FL
Supplier:  Amarell
Frequently Bought Together