ContactUsLinkComponent
Congealing point therm., similar DIN 12785, 0+100:0,5C, solid stem, white backed, red special liquid, total immersion, 300x6,5mm, durable pigment, null
Catalog # AMARM247300-FL/VC
Supplier:  Amarell
Congealing point therm., similar DIN 12785, 0+100:0,5C, solid stem, white backed, red special liquid, total immersion, 300x6,5mm, durable pigment, null
Catalog # AMARM247300-FL/VC
Supplier:  Amarell
Frequently Bought Together