ContactUsLinkComponent
Therm. straight stem, similar DIN 16174, -100+30:1C, cap. prismatic, yellow backed, milk glass scale, toluene filled, upper part 23 5x20,5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
  AMART91020-TOL
 :  Amarell
Therm. straight stem, similar DIN 16174, -100+30:1C, cap. prismatic, yellow backed, milk glass scale, toluene filled, upper part 23 5x20,5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
  AMART91020-TOL
 :  Amarell
Frequently Bought Together