ContactUsLinkComponent
Glass therm., industrial, V-shape, DIN 16181, -60+40:2C, type B, built-in length 63mm, special liquid filled, upper part 110mm, sma ll straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART123000-FL
Supplier:  Amarell
Glass therm., industrial, V-shape, DIN 16181, -60+40:2C, type B, built-in length 63mm, special liquid filled, upper part 110mm, sma ll straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART123000-FL
Supplier:  Amarell
Frequently Bought Together