ContactUsLinkComponent
Glass therm., industrial, V-shape, DIN 16189, 0+100:1C, built-in length 63mm, type B, special liquid filled, upper part 200mm, larg e straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART120044-FL
Supplier:  Amarell
Glass therm., industrial, V-shape, DIN 16189, 0+100:1C, built-in length 63mm, type B, special liquid filled, upper part 200mm, larg e straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART120044-FL
Supplier:  Amarell
Frequently Bought Together