ContactUsLinkComponent
Glass therm., industrial, V-shape, DIN 16189, -30+50:1C, built-in length 100mm, type B, special liquid filled, upper part 200mm, la rge straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART120024-FL
Supplier:  Amarell
Glass therm., industrial, V-shape, DIN 16189, -30+50:1C, built-in length 100mm, type B, special liquid filled, upper part 200mm, la rge straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART120024-FL
Supplier:  Amarell
Frequently Bought Together