ContactUsLinkComponent
Bimetal therm., tubular surface, 0+120:2C, diameter of case 80mm, to fit tubes of 1 to 2 in diameter 1 * 1 items
Catalog # AMART111908
Supplier:  Amarell
Bimetal therm., tubular surface, 0+120:2C, diameter of case 80mm, to fit tubes of 1 to 2 in diameter 1 * 1 items
Catalog # AMART111908
Supplier:  Amarell
Frequently Bought Together