ContactUsLinkComponent
Refill AMA Freezer therm., -35+20:0,5C, c. 250x17mm, enclosed scale, cap. yellow backed, red alcohol filled, rounded top finish, g overnment verified with Verification Stamp null
Catalog # AMART108420-FL/VS
Supplier:  Amarell
Refill AMA Freezer therm., -35+20:0,5C, c. 250x17mm, enclosed scale, cap. yellow backed, red alcohol filled, rounded top finish, g overnment verified with Verification Stamp null
Catalog # AMART108420-FL/VS
Supplier:  Amarell
Frequently Bought Together