ContactUsLinkComponent
Quartz-Glass therm., 0+750:2C, 450x8-9mm, cap. colourless, gallium filled, strip bearing the scale: quartz, total immersion, pin-to p finish with button, suitable for government verification 1 * 1 items
Catalog # AMARQ134000-GA
Supplier:  Amarell
Quartz-Glass therm., 0+750:2C, 450x8-9mm, cap. colourless, gallium filled, strip bearing the scale: quartz, total immersion, pin-to p finish with button, suitable for government verification 1 * 1 items
Catalog # AMARQ134000-GA
Supplier:  Amarell
Frequently Bought Together