ContactUsLinkComponent
Therm. to set Amarell Universal, +350+402:0,1C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersi on, richter-top finish, null
Catalog # AMARN71020-GA/WC
Supplier:  Amarell
Therm. to set Amarell Universal, +350+402:0,1C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersi on, richter-top finish, null
Catalog # AMARN71020-GA/WC
Supplier:  Amarell
Frequently Bought Together