ContactUsLinkComponent
Therm. to set Amarell Universal, +147+202:0,1C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersio n, richter-top finish, government calibrated with Calibration Certificate with 3 test points 1 * 1 items
Catalog # AMARN71012-GA/CC
Supplier:  Amarell
Therm. to set Amarell Universal, +147+202:0,1C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersio n, richter-top finish, government calibrated with Calibration Certificate with 3 test points 1 * 1 items
Catalog # AMARN71012-GA/CC
Supplier:  Amarell
Frequently Bought Together