ContactUsLinkComponent
Therm. to set Amarell Universal, -100+30:0,5C, 400x9mm, enclosed scale, cap.lens front, colourless, toluene, total immersion, richt er-top finish, suitable for government verification 1 * 1 items
Catalog # AMARN71002-TOL
Supplier:  Amarell
Therm. to set Amarell Universal, -100+30:0,5C, 400x9mm, enclosed scale, cap.lens front, colourless, toluene, total immersion, richt er-top finish, suitable for government verification 1 * 1 items
Catalog # AMARN71002-TOL
Supplier:  Amarell
Frequently Bought Together