Precision therm., similar ISO 656, short, -55+2:0,5C, milk-glass scale, cap. colourless, toluene filled, total immersion, max. 250x 9mm, richter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARN65780-TOL
Supplier: Amarell
Frequently Bought Together