ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 3d, solid stem white backed, 0+120:0,5C, length 430mm, imm. 100mm, capillary tube specially co ated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400010-CB
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 3d, solid stem white backed, 0+120:0,5C, length 430mm, imm. 100mm, capillary tube specially co ated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400010-CB
Supplier:  Amarell
Frequently Bought Together