ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 1d, solid stem, white backed, -10+20:0.,1C, length 410mm, imm. 100mm, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400000-CB
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 1d, solid stem, white backed, -10+20:0.,1C, length 410mm, imm. 100mm, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400000-CB
Supplier:  Amarell
Frequently Bought Together