ContactUsLinkComponent
Thermometer similar to IP 78 C, solid stem, white backed, 24+78:0,2C, capillary tube specially coated inside, with non-wetting blu e special liquid, immersion 100mm, 395x6-8mm, durable pigment, null
Catalog # AMARA341090-CB
Supplier:  Amarell
Thermometer similar to IP 78 C, solid stem, white backed, 24+78:0,2C, capillary tube specially coated inside, with non-wetting blu e special liquid, immersion 100mm, 395x6-8mm, durable pigment, null
Catalog # AMARA341090-CB
Supplier:  Amarell
Frequently Bought Together