ContactUsLinkComponent
Thermometer similar to IP 75 C, solid stem, white backed, -30+80:0,5C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
  AMARA341060-FL
 :  Amarell
Thermometer similar to IP 75 C, solid stem, white backed, -30+80:0,5C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
  AMARA341060-FL
 :  Amarell
Frequently Bought Together