ContactUsLinkComponent
Thermometer similar to IP 75 C, solid stem, white backed, -30+80:0,5C, capillary tube specially coated inside, with non-wetting bl ue special liquid, durable pigment, null
Catalog # AMARA341060-CB
Supplier:  Amarell
Thermometer similar to IP 75 C, solid stem, white backed, -30+80:0,5C, capillary tube specially coated inside, with non-wetting bl ue special liquid, durable pigment, null
Catalog # AMARA341060-CB
Supplier:  Amarell
Frequently Bought Together