ContactUsLinkComponent
Thermometer similar to IP 67 C, solid stem, white backed, -19,4-16,6:0,05C, capillary tube specially coated inside, with non-wetti ng blue special liquid, total immersion, 305x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA340920-CB
Supplier:  Amarell
Thermometer similar to IP 67 C, solid stem, white backed, -19,4-16,6:0,05C, capillary tube specially coated inside, with non-wetti ng blue special liquid, total immersion, 305x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA340920-CB
Supplier:  Amarell
Frequently Bought Together