ContactUsLinkComponent
Viscometer therm., similar Engler, similar DIN 12785, 0+105:0,5C, enclosed scale, cap. prismatic, colourless, red special liquid, 3 00x6mm, 90mm immersion, with swelling, Richter-top finish, without button, suitable for government verification 1 * 1 items
Catalog # AMARM238042-FL
Supplier:  Amarell
Viscometer therm., similar Engler, similar DIN 12785, 0+105:0,5C, enclosed scale, cap. prismatic, colourless, red special liquid, 3 00x6mm, 90mm immersion, with swelling, Richter-top finish, without button, suitable for government verification 1 * 1 items
Catalog # AMARM238042-FL
Supplier:  Amarell
Frequently Bought Together