ContactUsLinkComponent
Hydrometer DIN 12791/BS 718, M50, 0,80-0,85:0,0010g/cm , ref. temp. 20C, without therm., max. 270mm long, suitable for government v erification 1 * 1 items
Catalog # AMARH808008
Supplier:  Amarell
Hydrometer DIN 12791/BS 718, M50, 0,80-0,85:0,0010g/cm , ref. temp. 20C, without therm., max. 270mm long, suitable for government v erification 1 * 1 items
Catalog # AMARH808008
Supplier:  Amarell
Frequently Bought Together