ContactUsLinkComponent
Hydrometer DIN 12791/BS 718, L50, 0,85-0,90:0,0005g/cm , ref. temp. 20C, without therm., max. 335mm long, suitable for government v erification 1 * 1 items
Catalog # AMARH806210
Supplier:  Amarell
Hydrometer DIN 12791/BS 718, L50, 0,85-0,90:0,0005g/cm , ref. temp. 20C, without therm., max. 335mm long, suitable for government v erification 1 * 1 items
Catalog # AMARH806210
Supplier:  Amarell
Frequently Bought Together