ContactUsLinkComponent
Thermometer similar to IP 1 C, solid stem, white backed, -38+50:1C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 108mm, 230x6-8mm, durable pigment, null
Catalog # AMARA340000-CB
Supplier:  Amarell
Thermometer similar to IP 1 C, solid stem, white backed, -38+50:1C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 108mm, 230x6-8mm, durable pigment, null
Catalog # AMARA340000-CB
Supplier:  Amarell
Frequently Bought Together