ContactUsLinkComponent
Well scoop therm., -5+35:0,2C, milk glass scale, cap. prismatic, colourless, capillary tube specially coated inside, with non-wett ing blue special liquid, total immersion, in slotted brass armour with upper part c.200x20mm and scoop vessel c.70x32 1 * 1 items
Catalog # AMARK196120-CB/CC
Supplier:  Amarell
Well scoop therm., -5+35:0,2C, milk glass scale, cap. prismatic, colourless, capillary tube specially coated inside, with non-wett ing blue special liquid, total immersion, in slotted brass armour with upper part c.200x20mm and scoop vessel c.70x32 1 * 1 items
Catalog # AMARK196120-CB/CC
Supplier:  Amarell
Frequently Bought Together