ContactUsLinkComponent
Density hydrometer, 1,700-1,800:0,001g/cm , ref. temp. 20C, with therm., 0+40:1C, 350mm long, special liquid 1 * 1 items
Catalog # AMARH801222-FL
Supplier:  Amarell
Density hydrometer, 1,700-1,800:0,001g/cm , ref. temp. 20C, with therm., 0+40:1C, 350mm long, special liquid 1 * 1 items
Catalog # AMARH801222-FL
Supplier:  Amarell
Frequently Bought Together