Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Density hydrometer, 1,100-1,200:0,001g/cm , ref. temp. 20C, with therm., 0+40:1C, 350mm long, special liquid 1 * 1 items
Catalog # AMARH801210-FL
Supplier:
Amarell
Expand image
Density hydrometer, 1,100-1,200:0,001g/cm , ref. temp. 20C, with therm., 0+40:1C, 350mm long, special liquid 1 * 1 items
Catalog # AMARH801210-FL
Supplier:
Amarell
Frequently Bought Together
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Potassium fluoride ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Calibration set (includes 800, 100, 20.0 <(>&<)> 0.02 NTU standard) 1 * 1 items
Supplier: VELP SCIENTIFIC