ContactUsLinkComponent
Precision therm., straight stem, -20+150:1C, cap. lens front, colourless, special liquie, upper part 275x11mm, lower part 200x8mm, null
Catalog # AMARL36736-FL/VC
Supplier:  Amarell
Precision therm., straight stem, -20+150:1C, cap. lens front, colourless, special liquie, upper part 275x11mm, lower part 200x8mm, null
Catalog # AMARL36736-FL/VC
Supplier:  Amarell
Frequently Bought Together