ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 99 C, white backed, -50+5:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 35mm, max.305x6,5-8mm, null
Catalog # AMARA301368-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 99 C, white backed, -50+5:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 35mm, max.305x6,5-8mm, null
Catalog # AMARA301368-CB
Supplier:  Amarell
Frequently Bought Together