ContactUsLinkComponent
Refill thermometer for depth thermometer, similar DIN 58664, -10+30:0,1C, milk glass scale, capillary tube specially coated inside , with non-wetting blue special liquid, max.330x16mm, total immersion, null
Catalog # AMARK188448-CB/CC
Supplier:  Amarell
Refill thermometer for depth thermometer, similar DIN 58664, -10+30:0,1C, milk glass scale, capillary tube specially coated inside , with non-wetting blue special liquid, max.330x16mm, total immersion, null
Catalog # AMARK188448-CB/CC
Supplier:  Amarell
Frequently Bought Together