ContactUsLinkComponent
In-Door Psychrometer, lacquered wooden base c.250x120mm, water basin, 2 therm. -10+60:0,5C, milk glass scale, cap. prismatic, colou rless, red special liquid, with table for determination of relative humidity, suitable for government verification 1 * 1 items
Catalog # AMARK175340-FL
Supplier:  Amarell
In-Door Psychrometer, lacquered wooden base c.250x120mm, water basin, 2 therm. -10+60:0,5C, milk glass scale, cap. prismatic, colou rless, red special liquid, with table for determination of relative humidity, suitable for government verification 1 * 1 items
Catalog # AMARK175340-FL
Supplier:  Amarell
Frequently Bought Together